Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hafnium(IV) oxide, tech.

Click to view available options
Quantity:
100g
500g
12055-23-1
MFCD00003565
12055-23-1
MFCD00003565
Merck Index 14,4588
Solubility Information Insoluble in water.
Formula Weight 210.49
Physical Form -325 Mesh Powder
Grade Technical
Density 9.68 g/mL
Mesh Size −325 Mesh
Odor Odorless
Melting Point 2774°C
Quantity 500g
Chemical Name or Material Hafnium(IV) oxide
Show More Show Less

EINECSNumber : 235-013-2

TSCA : Yes

Recommended Storage : Ambient temperatures