Alfa Aesar™ Hafnium(IV) sulfate, 99.9% (metals basis excluding Zr), Zr <1%

Manufacturer: Alfa Aesar™ 4424406

Quantity 5g

 View more versions of this product

Catalog No. AA4424406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15823-43-5
    Molecular Formula Hf(SO4)2
    Formula Weight 370.62 (Anhydrous)
    MDL Number MFCD00168081
    Chemical Name or Material Hafnium (IV) sulfate

    Specifications

    Formula Weight 370.62 (Anhydrous)
    Physical Form Powder
    CAS 15823-43-5
    TSCA TSCA
    Molecular Formula Hf(SO4)2
    MDL Number MFCD00168081
    Quantity 5g
    Chemical Name or Material Hafnium (IV) sulfate
    Assay 99.9%, (metals basis excluding Zr), Zr <1%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit