Alfa Aesar™ Hafnium powder, -100 mesh, 99.8% (metals basis excluding Zr), Zr nominal <4.5%

Manufacturer: Alfa Aesar™ 7774922

Quantity 100g

 View more versions of this product

Catalog No. AA7774922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-58-6
    MDL Number MFCD00011032
    UN Number UN2545
    Chemical Name or Material Hafnium

    Specifications

    MDL Number MFCD00011032
    Health Hazard 2 GHS H Statement
    H251
    Self-heating
    may catch fire.
    Health Hazard 3 GHS P Statement
    P280-P235+P410-P420-P407
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep cool.
    Protect from sunlight.
    Store away from other materials.
    Maintain air gap between stacks/pallets.
    TSCA TSCA
    CAS 7440-58-6
    UN Number UN2545
    Mesh Size −100 mesh
    Odor Odorless
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.2; Packaging Group: II; Proper Shipping Name: HAFNIUM POWDER, DRY
    RTECS Number MG4600000
    Quantity 100g
    Chemical Name or Material Hafnium
    Assay 99.8%, (metals basis excluding Zr), Zr nominal <4.5%

    GHS H Statement
    H251
    Self-heating
    may catch fire.

    GHS P Statement
    P280-P235+P410-P420-P407
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep cool.
    Protect from sunlight.
    Store away from other materials.
    Maintain air gap between stacks/pallets.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit