Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hafnium powder, -60+325 mesh, 99.6% (metals basis excluding Zr), Zr nominal 2-3.5%

Manufacturer:  Alfa Aesar™ 1020204

Quantity 2g

 View more versions of this product

Catalog No. AA1020204

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-58-6
MFCD00011032
UN2545
Hafnium powder

Specifications

2g
Powder
UN2545
Odorless
Hafnium powder
7440-58-6
MFCD00011032
−60+325 Mesh
Gray
99.6%, (metals basis excluding Zr), Zr nominal 2 to 3.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H251
Self-heating: may catch fire.

GHS P Statement
P280-P235+P410-P420-P407
Wear protective gloves/protective clothing/eye protection/face protection.
Keep cool.
Protect from sunlight.
Store away from other materials.
Maintain air gap between stacks/pallets.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: HAFNIUM POWDER, DRY

RTECSNumber : MG4600000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit