Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium sponge, 99.6% (metals basis excluding Zr), Thermo Scientific™

Catalog Number AA0067430
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
7440-58-6
Hf
178.49
MFCD00011032
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
23986
CHEBI:33343
hafnium
[Hf]
7440-58-6
Hf
178.49
MFCD00011032
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
23986
CHEBI:33343
hafnium
[Hf]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver gray sponge
Assay Percent Range 99.6%, Zr nominal 3% (Metals basis excluding Zr)
Quantity 250 g
Odor Odorless
Form Sponge
Chemical Name or Material Hafnium
Hazard Category H250
Precautionary Statement P210-P222-P280-P335+P334-P370+P378q
DOTInformation DOT Class: 4.2, Packaging Group: II
EINECSNumber 231-166-4
TSCA Yes
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit