Hafnium sponge, 99.6% (metals basis excluding Zr), Alfa Aesar™

Manufacturer: Alfa Aesar 0067409

Quantity 10g

 View more versions of this product

Catalog No. AA0067409

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-58-6
    Physical Form Sponge, 0.8-20mm (0.03-0.79 in.)
    Assay Percent Range 99.6%, Zr nominal 3% (Metals basis excluding Zr)
    MDL Number MFCD00011032
    Chemical Name or Material Hafnium

    Specifications

    Quantity 10g
    Color Silver-gray
    DOT Information DOT Class: 4.2, Packaging Group: II
    EINECS Number 231-166-4
    Physical Form Sponge, 0.8-20mm (0.03-0.79 in.)
    Assay Percent Range 99.6%, Zr nominal 3% (Metals basis excluding Zr)
    MDL Number MFCD00011032
    CAS 7440-58-6
    Odor Odorless
    Health Hazard 1 H251: Self-heating: may catch fire.
    Appearance Silver gray sponge
    Chemical Name or Material Hafnium

    H251: Self-heating: may catch fire.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit