Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hafnium tert-butoxide, 99.9% (metals basis excluding Zr), Zr< 0.5%

Manufacturer:  Alfa Aesar™ 4190804

Quantity 2g

 View more versions of this product

Catalog No. AA4190804


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2172-02-3
470.94
UN1993
Hf[OC(CH3)3]4
MFCD00070458
Hafnium tert-butoxide

Specifications

28° (82°F)
90°C (5mmHg)
2°C
2172-02-3
Liquid
470.94
UN1993
Hafnium tert-butoxide
Air & Moisture Sensitive
Pale yellow
2g
1.15g/mL
Hf[OC(CH3)3]4
MFCD00070458
Soluble in hydrocarbons, reacts with alcohols, ketones, and esters
99.9%, (metals basis excluding Zr), Zr < 0.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit