Alfa Aesar™ Hafnium wire, 2.0mm (0.08 in.) dia., 99.7% (metals basis excluding Zr), Zr nominal 3%

Manufacturer: Alfa Aesar™ 13305CC

Quantity 500cm

 View more versions of this product

Catalog No. AA13305CC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-58-6
    MDL Number MFCD00011032
    Chemical Name or Material Hafnium wire, 2.0mm (0.08 in.) dia.

    Specifications

    Physical Form Wire
    CAS 7440-58-6
    Odor Odorless
    Weight ≈41.1g/m
    Quantity 500cm
    TSCA TSCA
    MDL Number MFCD00011032
    Chemical Name or Material Hafnium wire, 2.0mm (0.08 in.) dia.
    Assay 99.7%, (metals basis excluding Zr), Zr nominal 3%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit