Learn More
Abnova™ HAGH HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Human overexpression lysate
Manufacturer:  Abnova™ L123T6
Catalog No. 89-947-066
Add to cart
Specifications
Specifications
HEK293
NM_005326
Theoretical M.W. 29kDa
Transient overexpression cell lysate was tested with anti-HAGH antibody (Mfr. No. H00003029-M02A) by Western Blot.
In modified RIPA Lysis Buffer.
Human
Immunoprecipitation, Western Blot
HAGH HEK293 Cell Transient Overexpression Lysate(Non-Denatured)
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
pCMV-HAGH full length
100 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
2 mg/ml