Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAGHL Mouse anti-Human, Mouse, Rat, Mammalia, PE/Atto594, Clone: 8.IGSH, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP197479PEATT594

Catalog No. N197479P594

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HAGHL Monoclonal antibody specifically detects HAGHL in Human, Mouse, Rat, Mammalia samples. It is validated for Flow Cytometry.
Specifications

Specifications

HAGHL
8.IGSH
Flow Cytometry
Conjugated, Purified
HAGHL
Thiol-modified glutathione conjugated to KLH.
Protein A or G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mammalia, Mouse, Rat
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
EC 3.1.2.-, GLO2-like/ RJD12, hydroxyacyl glutathione hydrolase-like, hydroxyacylglutathione hydrolase-like, hydroxyacylglutathione hydrolase-like protein, MGC2605
Mouse
IgG1
0.1 mL
Primary
84264.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit