Learn More
HAI-1/HGFA Inhibitor 1 Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0088-3G21, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212356AF594
Catalog No. NP21236A94
Add to cart
Description
Description
HAI-1/HGFA Inhibitor 1 Monoclonal antibody specifically detects HAI-1/HGFA Inhibitor 1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
HAI-1/HGFA Inhibitor 1
RM0088-3G21
Western Blot
HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
SPINT1
Mouse HGFA Inhibitor 1
0.1 mL
Primary
6692