Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HAI-1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552361
Description
Immunogen sequence: ACFLINCLYE QNFVCKFAPR EGFINYLTRE VYRSYRQLRT QGFGGSGIPK AWAGIDLKVQ PQEPLVLKDV ENTDWRLLRG DTDVRVERKD PNQVELWGLK EGTYLFQLTV TSSDHP Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 73%.
The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms.
Specifications
HAI-1 | |
Polyclonal | |
Unconjugated | |
SPINT1 | |
HAI; HAI1; HAI-1; hepatocyte growth factor activator inhibitor 1; hepatocyte growth factor activator inhibitor type 1; hepatocyte growth factor activator inhibitor-1; kunitz-type protease inhibitor 1; MANSC2; serine peptidase inhibitor, Kunitz type 1; serine protease inhibitor, Kunitz type 1; Spint1; UNQ223/PRO256 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6692 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43278 | |
SPINT1 | |
Recombinant protein corresponding to Human HAI-1. Recombinant protein control fragment (Product #RP-88700). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction