Learn More
HAI-1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150361AP
Catalog No. 50-172-9921
Add to cart
Description
Description
The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms.
Specifications
Specifications
HAI-1
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43278, Q9R097
HAI, HAI 1, HAI1, MANSC2, SPINT1
Rabbit
IgG
150 μL
-20°C
Primary
20732, 6692
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SPINT1
Liquid
SPINT1
HAI-1 Fusion Protein Ag6665
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse