Learn More
HAI-2/SPINT2 Rat anti-Mouse, DyLight 488, Clone: RM0089-7F52, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP121728G
Catalog No. NBP121728G
Add to cart
Description
Description
HAI-2/SPINT2 Monoclonal antibody specifically detects HAI-2/SPINT2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
HAI-2/SPINT2
RM0089-7F52
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
SPINT2
Mouse recombinant of the common extracellular region of HAI-2 isoforms
Protein G purified
Cancer
Primary
10653
Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the common extracellular region of HAI-2 isoforms.