Learn More
Sklar InstrumentsSupplier Diversity Partner SklarLite Sterilization Half Size Container, Sklar, 11 3/4 in. x 11 in. x 6 in. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 104596
SklarLite Sterilization Half Size Container, Sklar, 11 3/4 in. x 11 in. x 6 in., Non-Perforated Bottom, Silver, Anodized Aluminum Alloy. SklarLite Sterilization Containers are made from anodized aluminum that allow them to be durable but lighter than traditional containers, ideal for not only sterilization and storage, but also easy transport. Available in multiple sizes, with lids available in a variety of colors, containers are available in sets or individual components. Sklar also offers a wide range of accessories, including baskets, silicone mats, custom ID labels, and filters. This product is a half size container with a non-perforated bottom and a silver lid.
Catalog No. 50-143-7192