Learn More
Alfa Aesar™ Haloenol Lactone Suicide Substrate, ≥98%
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J63728MA
 View more versions of this product
Catalog No. AAJ63728MA
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
White
Lyophilized
10mg
Irreversibly inhibits calcium-independent phospholipase A-2
317.18
-30° to -10°C