Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Alfa Aesar™ Haloenol Lactone Suicide Substrate, ≥98%
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J63728MB
 View more versions of this product
Catalog No. AAJ63728MB
Order in Bulk
Request bulk or custom formats
Specifications
Specifications
White
Lyophilized
25mg
Irreversibly inhibits calcium-independent phospholipase A-2
317.18
-30° to -10°C