Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Halogen Moisture Analyzer

Mettler Toledo™ Halogen Moisture Analyzer is designed to provide high performance moisture analysis.

$9215.00 - $11720.00

Specifications

Display WVGA 7 in. Touch Screen Color Display
Temperature Range (English) 104 to 446°F
Temperature Range (Metric) 40 to 230°C
Type Halogen Moisture Analyzer
Repeatability Automatic (5), Timed, Free
View More Specs

Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
13-940-093 View Documents Mettler Toledo™
30019875
HX204 (Internal Calibration) Each for $11,720.00
Add to cart
 
13-940-094 View Documents Mettler Toledo™
30019876
HS153 (External Calibration) Each for $9,215.00
Add to cart
 
Description

Description

  • Accurate results
  • Optimized productivity
  • OneClick™ Operation
  • Safety ensuring user management
  • Built in performance assurance tests
Specifications

Specifications

WVGA 7 in. Touch Screen Color Display
40 to 230°C
Automatic (5), Timed, Free
104 to 446°F
Halogen Moisture Analyzer
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit