Learn More
Medchemexpress LLC Haloperidol (standard) | 52-86-8 | 99.8% | C₂₁H₂₃ClFNO₂ | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14538R
Encompass_Preferred
Haloperidol (Standard) is the analytical standard of Haloperidol, primarily intended for research and analytical applications. It acts as a potent dopamine D2 receptor antagonist, widely recognized as an antipsychotic.
  • Analytical standard for Haloperidol
  • Potent dopamine D2 receptor antagonist
  • Widely used as an antipsychotic
  • Reference standard supplied assay
  • Commonly used in HPLC, GC, and MS experiments
Catalog No. 50-003-55154