Learn More
Promega HaloTag™ Cloning Starter System
HaloTag™ Cloning Starter Kit: designed to use Flexi Cloning System to clone protein coding sequences into mammalian HaloTag™ expression vectors; clone and transfer gene sequences into C- or N-terminus of HaloTag™ Flexi CMV Deletion Vectors.
Supplier:  Promega G6050
Catalog No. PRG6050
Add to Cart
Includes: Wizard™ SV Gel and PCR Clean-Up System
Flexi™ System, Entry/Transfer
HaloTag™ Flexi™ Vectors - CMV Deletion Series Sample Pack
Carboxy Flexi™ Enzyme Blend (SgfI & EcoICRI)
Description
Description
This series of vectors containing deletions of human cytomegalovirus (CMV) immediate-early enhancer/promoter were created to provide different constitutive expression levels of HaloTag™ 7 fusion proteins, providing greater control over the level of protein expression. This allows expression of enough fusion protein to image or capture but not so much to disturb normal function of the protein partner fused to the HaloTag™ 7 protein.
Specifications
Specifications
Components at different temps
1/Ea.
Product Suggestions
Product Suggestions