Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Halsey Needle Holder, Sklar, 5 in., Blunt, Straight, Smooth, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21810
Econo Halsey Needle Holder, Sklar, 5 in., Blunt, Straight, Smooth, Stainless Steel. Econo Halsey Needle Holders are used for holding small needles for suturing, most commonly during plastic surgery and procedures with delicate tissue, such as dermatology. They are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with smooth tips and a length of 5 inches. They are sold in boxes of 12.
Catalog No. 50-143-7781