Halyard Health BASIC™ Plus Lab Coats With Knit Collar and Cuffs
Protective tri-layer fabric offers resistance to blood strikethrough at various levels of exposure
Manufacturer: HALYARD HEALTH 10033H
 View more versions of this product
Catalog No. 23-500-167
  Check Availability
Add to Cart
    Specifications
    Material 3-layer SMS fabric
    Color Blue
    Size X-Large
    Size (Metric) Chest 72.35cm
    Length (Metric) 115.6cm
    Cuff Style Knit
  • Description & Specifications
Related Products
Product Content Feedback