Halyard™ Health Universal Precautions Lab Jackets

Protective tri-layer fabric offers resistance to blood strikethrough at various levels of exposure

Manufacturer: HALYARD HEALTH 10072H

 View more versions of this product

Catalog No. 23-500-188

  Check Availability
Add to Cart

    Specifications

    No. of Pockets 3 reinforced
    Collar Type Knit
    Cuff Style Knit
    Color White
    Size Large
    Length (Metric) Sleeve 49.5cm
    Size (Metric) Chest 68.24cm
    Sterile No
    Product Type Universal Precautions Lab Jacket
    Length (Metric) Back: 77.5cm
    Material 3-layer SMS

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit