Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Halyard™ Health Universal Precautions Lab Jackets

Protective tri-layer fabric offers resistance to blood strikethrough at various levels of exposure

Manufacturer:  HALYARD HEALTH 10071H

 View more versions of this product

Catalog No. 23-500-187


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

3-layer SMS; Reinforced pockets
3
Collared
Knit
Medium
49.5cm
Universal Precautions Lab Jacket
SMS
60.75cm
Knit
White
Back: 74.9cm
No
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit