Learn More
O&M Halyard Quick Check Sterilization Wrap (H300)
Using Halyard Quick Check wraps is a fast, easy way to ensure instrument sterility is uncompromised.
$162.77 - $464.34
Specifications
Certifications/Compliance Meets NFPA Class 1
Sterility Non-sterile
Latex Free Yes
Autoclavable Autoclavable
Color Blue
View More Specs
Products 8
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
23-500-350
View Documents
O&M
34171
60 x 60 cm (24 x 24 in.)
Case of 240 for $270.97
Add to cart
23-500-351
View Documents
O&M
34173
76 x 76 cm (30 x 30 in.)
Case of 144 for $253.97
Add to cart
23-500-352
View Documents
O&M
34160
91 x 91 cm (36 x 36 in.)
Case of 144 for $395.49
Add to cart
23-500-353
View Documents
O&M
34189
101 x 101 cm (40 x 40 in.)
Case of 96 for $285.54
Add to cart
23-500-354
View Documents
O&M
34159
114 x 114 cm (45 x 45 in.)
Case of 48 for $162.77
Add to cart
23-500-355
View Documents
O&M
34181
121 x 121 cm (48 x 48 in.)
Case of 96 for $464.34
Add to cart
23-500-356
View Documents
O&M
34182
137 x 137 cm (54 x 54 in.)
Case of 48 for $235.43
Add to cart
23-500-357
View Documents
O&M
34190
137 x 182 cm (54 x 72 in.)
Case of 48 for $314.29
Add to cart
Description
Description
  • Inspect trays easier and faster than ever
  • Superior microbial barrier protection with patented Powerguard technology
  • Cleared for all of the same sterilization systems as Halyard Kimguard One-step Wrap
  • Made from recyclable No. 5 polypropylene fabric
  • Made in the USA
Specifications
Specifications
Meets NFPA Class 1
Yes
Blue
Non-sterile
Autoclavable
Basin sets, instrument sets, procedure trays and large linen packs (6 to 9 lb.)