Learn More
Hamilton™ 1700/1000 Series Gastight™ Instrument Syringes: AD Termination
Syringes are designed to fit into, or with, various different instruments such as the MICROLAB™ 500 and MICROLAB 1000
Manufacturer:  Hamilton™ 201250
 View more versions of this product
Catalog No. 13-684-79P
  Check Availability
Add to cart
Specifications
Description
Have a PTFE plunger tip that creates a tight connection between the syringe barrel and plunger allowing for liquid or gas sampling.
  • AccuDil™ (AD) - This termination is designed for MICROLAB 1000 Diluters/Dispensers
Description & Specifications
Specifications
For Microlab™ 1000 diluters/dispensers
No Needle Included
PTFE
122°F
68°F
No Needle Included
Each
Gastight™ Syringe
2.5mL
No Needle Included
Borosilicate Glass
200psi
50°C
1002
N/A
AccuDil™
No needle included