Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamilton™ 1700/1000 Series Gastight™ Instrument Syringes: CX Termination

Syringes are designed to fit into, or with, various different instruments such as the MICROLAB™ 500 and MICROLAB 1000

Manufacturer:  Hamilton™ 81162

 View more versions of this product

Catalog No. 14-815-360


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Yes
0.305 in.
7.75mm
Instrument plunger stop
No Needle Included
PTFE
239°F
50°F
No Needle Included
Each
Gastight™ Syringe
250μL
0.091 in.
2.3mm
For dispensing volumes from 100μL up to 100mL
No Needle Included
Borosilicate Glass
500psi
115°C
1725
N/A
ChemSeal
No needle included
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit