Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamilton™ 1700 Series Gastight™ Gel Loading Syringes, 10uL

Syringes are designed to work for multiplex DNA sequencing applications

$109.40

Specifications

Autoclavable Not Autoclavable
Volume (Metric) 10 µL
For Use With (Application) Designed to work for multiplex DNA sequencing applications
Max. Pressure 1000 psi
Max. Temperature (English) 122°F
View More Specs

Products 1
Catalog Number Mfr. No. Diameter (Metric) Outer Needle Type Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Outer Needle Type Length (English) Length (Metric) Price Quantity & Availability  
14-815-365
View Documents
Hamilton™
80081
0.2 mm Single-Channel Gel Loading Syringe 2 in. 5.08 cm
Each for $109.40
Add to cart
 
Description

Description

GASTIGHT syringes with a PTFE plunger tip that creates a tight connection between the syringe barrel and plunger allowing for liquid or gas sampling

  • Cemented Needle (N) Termination: For low volume syringes, the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable
  • Syringe accurately dispenses from 0.2 to 10μL of sample per syringe simultaneously
  • Point style 3: Blunt needle point for use with HPLC injection valves and for sample pipetting
  • Available gauges: 0.2, 0.3, 0.4 mm
  • 10μL volume

Specifications

Specifications

Not Autoclavable
Designed to work for multiplex DNA sequencing applications
122°F
3
10 µL
1000 psi
50°C
Sterile
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit