Hamilton™ 1700 Series Gastight™ Gel Loading Syringes: RN Termination, 2.75 in. (70mm) Needle Length: RN Termination, 2.75 in. (70mm) Needle Length
Syringes are designed to work for multiplex DNA sequencing applications
Manufacturer: Hamilton™ 78633
 View more versions of this product
Catalog No. 14-815-322
  Check Availability
Add to Cart
    Specifications
    Autoclavable Yes
    For Use With (Application) Designed to work for multiplex DNA sequencing applications
    Needle Point Number 3
    Length (English) 2.75 in.
    Length (Metric) 6.9cm
    Needle Gauge 0.2mm
    Product Type Replacement Needle
  • Description & Specifications
Related Products
Product Content Feedback