Hamilton™ 1800 Series Gastight™ Replacement Syringe Barrels
Manufacturer: Hamilton™ 32189
 View more versions of this product
Catalog No. 14-819-275
  Check Availability
Add to Cart
    Specifications
    Material PTFE
    Volume (Metric) 50μL
    Volume (English) 0.00169 oz.
    Type Replacement Syringe Barrel
    For Use With (Equipment) 1805 RN syringe
    For Use With (Application) Gases and liquids
  • Description & Specifications
Related Products
Product Content Feedback