Hamilton™ 7000 Series Gastight™ and Microliter™ Digital Syringes: KH Termination
Easily achieves precision up to 0.5% while dispensing volumes from 500µL down to 50nL
Manufacturer: Hamilton™ DS80100
 View all options for this product
Catalog No. 14-813-272
  Check Availability
Add to Cart
    Syringe eliminates parallax errors by providing a digital readout on its integrated LCD screen. An adjustable plunger-stop guarantees reproducible plunger placement.
    Read the syringe volume directly from the liquid crystal display. The digital display eliminates volume reading errors as well as ensures that everyone in the lab reads the syringe volume the same.
    Ideal for Instrument Calibration
    Every Digital Syringe is a calibrated device traceable to N.I.S.T. methods. Use it to calibrate analytical instruments (AA/ICP, GC, HPLC, etc.), prepare standards and spike samples. Provides easier to compliance with EPA, CLIA, ISO and FDA (GLP, GMP) requirements.
    Better Accuracy and Precision
    7000 series MODIFIED MICROLITER™ syringes are certified to ± 1.0% accuracy or better. Improved precision is easily achieved with the built-in adjustable plunger stop. The adjustable plunger stop (0-100% of syringe volume) is engaged with a touch of the button on the back of the digital syringe.
  • Description & Specifications
Related Products
Product Content Feedback