Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamilton™ 7000 Series Modified Microliter™ Syringes

The 7000 series is the only zero dead volume syringe, ideal for dispensing volumes from 0.05µL up to 5µL

Manufacturer:  Hamilton™ 88011

 View more versions of this product

Catalog No. 14-813-223


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Yes
0.012 in.
0.02 in.
0.32mm
0.56mm
2.75 in.
2000psi
115°C
10°C
Sharp, Bevel, Curved, Non-coring
Knurled hub
24 ga.
Removable
0.01 in.
0.3 in.
0.32mm
7.82mm
70mm
239°F
50°F
2
Each
General Purpose Syringe
5μL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit