Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamilton™ Constant Rate-700 HPLC Syringes & Replacement Parts

Syringes are designed to delive reasy, reproducible injection of liquid samples into an injector on HPLC instrumentation. The CR-700 standardizes three major variables: sample volume, rate of injection, and depth of needle penetration. Even when operated by different technicians, the CR-700 provides reproducibility of +1% of the total syringe volume in routine procedures.

Manufacturer:  Hamilton™ 84301

 View more versions of this product

Catalog No. 14-823A


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Gastight™ Syringe
22s ga
Blunt
0.0283 in.
0.72mm
2 in.
Borosilicate Glass
CR700-20
50° to 176°F
20μL
Each
0.0059 in.
0.15mm
HPLC injection valves
1000psi
51mm
Stainless Steel
10° to 80°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit