Learn More
Hamilton™ Gastight™ Spark Holland HPLC Autosampler Syringes
Feature rugged polytetrafluoroethylene (PTFE) plunger tip and are economically designed to offer long-lasting, top-level quality
Manufacturer:  Hamilton™ 5466201
 View more versions of this product
Catalog No. 13-621-734
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
0.029 in.
0.729mm
Gastight™ Autosampler Syringe
No Needle Included
No Needle Included
No Needle Included
0.4 in.
0.46cm
68.9 bar
50° to 239°F
2500μL
0.305 in.
7.75mm
1mL
Each
N/A
0.18 in.
1.03cm
Spark Holland midas triathlon, endurance, alias, integrity, optimas, varian prostar 410, prostar 420, prostar 430, dionex model 542, 540, 540MT, PerkinElmer™ series 225, FLEXAR, Beckman coulter 508, grace/alltech 580, antec AS 100, AS 11
1002C
10° to 115°C