Learn More
Hamilton™ Gastight™ Spark Holland HPLC Autosampler Syringes
Feature rugged polytetrafluoroethylene (PTFE) plunger tip and are economically designed to offer long-lasting, top-level quality
Manufacturer:  Hamilton™ 5465801
 View more versions of this product
Catalog No. 13-621-730
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
100μL
100μL
Each
N/A
No Needle Included
0.3 in.
0.14cm
68.9 bar
50° to 239°F
Gastight™ Autosampler Syringe
No Needle Included
No Needle Included
No Needle Included
0.057 in.
0.77cm
Spark Holland midas triathlon, endurance, alias, integrity, optimas, varian prostar 410, prostar 420, prostar 430, dionex model 542, 540, 540MT, PerkinElmer™ series 225, FLEXAR, Beckman coulter 508, grace/alltech 580, antec AS 100, AS 11
1710C
10° to 115°C