Learn More
Hamilton™ Gastight™ Waters WISP Autosampler Syringes
Designed for use with Waters WISP HPLC autosamplers
Manufacturer:  Hamilton™ 80020
 View more versions of this product
Catalog No. 14-815-18
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
0.305 in.
7.75mm
1000psi
115°C
10°C
Each
25μL
0.029 in.
0.72mm
Waters WISP HPLC autosamplers
239°F
50°F
1702C
Gastight™ Syringe