Learn More
Hamilton™ Gastight™ Waters WISP Autosampler Syringes
Designed for use with Waters WISP HPLC autosamplers
Manufacturer:  Hamilton™ 80024
 View more versions of this product
Catalog No. 14-815-100
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
0.305 in.
7.75mm
500psi
115°C
10°C
Each
250μL
0.091 in.
2.3mm
Waters WISP HPLC autosamplers
239°F
50°F
1725C
Gastight™ Syringe