Learn More
Hamilton™ Kel-F Hub Side Port Needles (Luer Lock)
Kel-F hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2 in. (51mm). Custom lengths and point styles are available.
Manufacturer:  Hamilton™ 774610
 View more versions of this product
Catalog No. 14-815-519
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.01 in.
0.26mm
Luer-tip and PTFE luer-lock syringes and connectors
51mm
Cone
26 ga.
0.01 in.
0.47mm
2 in.
Nickel Plated Brass
Kel-F Hub Needle