Learn More
Hamilton™ Metal Hub Blunt Point Needles (Luer Lock)
Can be used with TLL syringes and LT or TLL connectors.
Manufacturer:  Hamilton™ 91023
 View more versions of this product
Catalog No. 14-815-611
  Check Availability
Add to cart
Specifications
Description
  • Made of 304 Stainless Steel
  • Standard needle length is 51mm (2 in.)
  • Available in convenient six packs
  • Custom needle lengths and point styles are available
  • Needles are available from 10 to 33 ga
Description & Specifications
Specifications
Yes
0.025 in.
0.64mm
2 in.
Nickel Plated Brass
Blunt
23 ga.
0.013 in.
0.34mm
PTFE luer-lock syringes and luer-tip or PTFE luer-lock connectors
51mm
Stainless steel
Needle