Learn More
Hamilton™ Microlab 300 Pipetting System Accessories - Syringes
Choose from two syringes designed for use specifically with the Hamilton Microlab 300 Pipetting System
$112.10
Specifications
For Use With (Equipment) Microlab™ 300 pipetteting system
View More Specs
Products 2
Catalog Number Mfr. No. Includes Model Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Includes Model Volume (Metric) Price Quantity & Availability  
14-815-916
View Documents
Hamilton™
6608840
PTFE plunger tip 1002.5 1.25 μL
Each for $112.10
Add to cart
14-815-917
View Documents
Hamilton™
6608815
Plunger tip 1705.5 BFP 25 μL
Each for $112.10
Add to cart
Description
Description
Model 1705.5
  • Bubble Free Prime
  • Expels air from the syringe and valve thanks to an innovative plunger tip
  • 1.25mL
  • Model 1002.5
    • Gastight
    • Ideal for dispensing both liquids and gases
    • Has a precision-machined PTFE plunger tip which creates a leak-free seal
    • 25µL
Specifications
Specifications
Microlab™ 300 pipetteting system