Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Accessories - (BFP) Syringes
For use with MICROLAB 600 Diluter/Dispenser
$180.00 - $660.50
Specifications
For Use With (Equipment) Microlab™ 600 dispensers
View More Specs
Products 12
Catalog Number Mfr. No. Volume (Metric) Model Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Model Price Quantity & Availability  
14-816-269
View Documents
Hamilton™
5900005
10 μL 1701 BFP
Each for $180.00
Only null left
Add to Cart
14-816-271
View Documents
Hamilton™
5900015
50 μL 1705 BFP
Each for $186.40
Only null left
Add to Cart
14-816-272
View Documents
Hamilton™
5900020
100 μL 1710 BFP
Each for $188.50
Only null left
Add to Cart
14-816-270
View Documents
Hamilton™
5900010
25 μL 1702 BFP
Each for $188.60
Only null left
Add to Cart
14-816-273
View Documents
Hamilton™
5900025
250 μL 1725 BFP
Each for $188.00
Only null left
Add to Cart
14-816-274
View Documents
Hamilton™
5900030
500 μL 1750 BFP
Each for $206.00
Only null left
Add to Cart
14-816-275
View Documents
Hamilton™
5900035
1 mL 1001 BFP
Each for $216.00
Only null left
Add to Cart
14-816-276
View Documents
Hamilton™
5900040
2.5 mL 1002 BFP
Each for $228.00
Only null left
Add to Cart
14-816-278
View Documents
Hamilton™
5900050
10 mL 1010 BFP
Each for $317.50
Only null left
Add to Cart
14-816-280
View Documents
Hamilton™
5900060
50 mL Bubble free prime
Each for $660.50
Only null left
Add to Cart
14-816-279
View Documents
Hamilton™
5900055
25 mL Bubble free prime
Each for $570.50
Only null left
Add to Cart
14-816-277
View Documents
Hamilton™
5900045
5 mL 1005 BFP
Each for $251.00
Only null left
Add to Cart
Description
Description
Syringe has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Specifications
Specifications
Microlab™ 600 dispensers
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.