Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Accessories - (BFP) Syringes
For use with MICROLAB 600 Diluter/Dispenser
Manufacturer:  Hamilton™ 5900025
 View more versions of this product
Catalog No. 14-816-273
  Check Availability
Add to cart
Specifications
Description
Syringe has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Description & Specifications
Specifications
Microlab™ 600 dispensers
1725 BFP
250μL