Learn More
Hamilton™ Microlab™ Accessories - PTFE Tubing Assemblies
For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Supplier: Hamilton™ 117102
Description
- For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Specifications
FEP | |
3.75 μL/mm | |
1/4-28 Tubing threads |
Gauge 12, Dispense Tubing | |
Microlab™ 600 diluter/dispenser system |