Learn More
Hamilton™ Microlab™ Accessories - PTFE Tubing Assemblies
For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Manufacturer:  Hamilton™ 6161501
 View more versions of this product
Catalog No. 14-816-259
  Check Availability
Add to cart
Specifications
Description
  • For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Description & Specifications
Specifications
Microlab™ 600 diluter/dispenser system
47.99 in.
FEP
1.15mL
1/4-28 Tubing threads
1219mm
PTFE Tubing Assemblies