Hamilton™ PRP-C18 Reversed-Phase HPLC Column Accessories - Bulk Resin
Bulk resin for individuals who prefer to pack their own HPLC columns
Manufacturer: Hamilton™ 79791
 View more versions of this product
Catalog No. 13-622-119
  Check Availability
Add to Cart
    Specifications
    pH 1 to 14
    Material PRP-C18
    Particle Size 5μm
    Quantity 1g
    Max. Temperature >100°C
  • Description & Specifications
Related Products
Product Content Feedback