Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamilton™ PRP™-X200 Cation Exchange Columns

Analyze inorganic and organic cations using conductivity or UV detection. Separate mono or divalent cations depending on mobile phase conditions from 20 ppb to 200 ppm.

$776.00 - $984.00

Specifications

Column Format Analytical Column
Diameter (Metric) 4.1 mm
Column Type Ion Exchange
Particle Size 10 μm
Pore Size 100 Å
View More Specs

Products 2
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
14-816-15
View Documents
Hamilton™
79441
150 mm
Each for $776.00
Add to cart
 
14-816-16
View Documents
Hamilton™
79442
250 mm
Each for $984.00
Add to cart
 
Description

Description

Can be used with any existing HPLC or ion chromatograph to easily determine cations in air, water and soil. PRP-X200 columns provide a number of benefits

Better than Wet Chemical Methods

  • Ion chromatography will greatly reduce sample pretreatment and improve the accuracy and precision of results.
  • High Sensitivity
    • Separate ions at concentrations from 20ppb to 20ppm without suppression or elaborate preconcentration techniques
  • Long Column Life
    • Highly inert polymeric support resists chemical attack from organic solvents and aqueous buffers (0-100% aqueous or organic; pH 1 to 13), effectively lengthening column life. If column performance should deteriorate (peak broadening and a loss of symmetry), a regeneration protocol will usually return your chromatography to its original state.
  • Analyze Mono or Divalent Cations
    • Quickly and easily separate five monovalent cations in five minutes or four divalent cations in four minutes
  • Unique Selectivity
    • Highly inert poly(styrene-divinylbenzene) packing allows the use of organic modifier. By varying the concentration of methanol the resolution between cations can be altered.

    • One particle size: 10°m
    • Four column internal diameters: 2.1 to 10.0mm
    • Two column materials: 316 stainless steel and PEEK
    • Analytical and semiprep/preparative guard columns

    Specifications

    Specifications

    Analytical Column
    Ion Exchange
    100 Å
    Analyzing Inorganic and Organic Cations Using Conductivity or UV Detection
    5000 psi (345 bar)
    PSDVB/Sulfonic Acid
    5°C to 60°C
    L22
    4.1 mm
    10 μm
    2 mL/min
    Stainless Steel
    pH 1 to 13, 0 to 100% aqueous, organic modifier
    PRP-X200
    HPLC Column
    SDS
    Product Certifications
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit