Learn More
Hamilton™ Replacement Needles for Agilent 7673, 7683, 7693, and 6850 ALS GC Syringes
Can be used with removable (RN)-style syringes that are 100 µL and smaller. These needles are manufactured for use with Agilent Instrumentation 7673-7683, 6850, 7670, 7671, and 7672 ALS models. Custom lengths and point styles are available.
Manufacturer:  Hamilton™ 778501
 View more versions of this product
Catalog No. 14-815-543
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
1.69 in.
Removable AS
23s-26s ga
Agilent Instrumentation 7673, 7683, 7693, and 6850 ALS GC syringe
42.9mm
Replacement Needle