Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamilton™ Small Hub Removable Needles (2.5 - 100µL Syringes)

Can be used with removable (RN)-style syringes that are 100µL and smaller.

Manufacturer:  Hamilton™ 775804

 View more versions of this product

Catalog No. 14-815-538


  Check Availability
Add to cart

Specifications

Description

  • Standard needle length is 51mm (2 in.)
  • Custom gauges, lengths and point styles are available
Description & Specifications

Specifications

Yes
0.018 in.
0.46mm
2 in.
Stainless Steel
Replacement Syringe Needle
0.01 in.
0.26mm
Ideal when there is a risk of the needle clogging
51mm
Sharp, Bevel, Curved
26 ga.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit