Learn More
Hamilton Syringe 1.0uL from Restek
High precision and accuracy down to 0.1µL.
$61.00 - $120.00
Catalog Number | Mfr. No. | Volume (Metric) | Needle Gauge | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Needle Gauge | Price | Quantity & Availability | ||||
06-712-336
|
Restek Corporation![]() 24546 |
0.5 µL | 25 G |
Each for $112.00
|
|
06-712-337
|
Restek Corporation![]() 24547 |
1.0 µL | 25s G |
Each for $97.00
|
|
06-712-338
|
Restek Corporation![]() 24548 |
1.0 µL | 25s G |
Each for $94.72
|
|
06-712-335
|
Restek Corporation![]() 24545 |
0.5 µL | 25 G |
Each for $112.00
|
|
06-712-339
|
Restek Corporation![]() 24549 |
1.0 µL | 22s G |
Each for $94.00
|
|
06-712-345
|
Restek Corporation![]() 24555 |
5.0 µL | 24 G |
Each for $97.00
|
|
06-712-346
|
Restek Corporation![]() 24556 |
5.0 µL | 24 G |
Each for $120.00
|
|
06-712-367
|
Restek Corporation![]() 24577 |
1 mL | 22s G |
Each for $61.00
|
|
Description
- Sample is contained in the needle with full plunger displacement, ensuring no dead volume
- Seal between the plunger and the needle can be tightened for high-pressure injection
- Replaceable needle/plunger kits and barrels
- Needle Length: 70mm (2.75 in.)
Specifications
Hamilton Micro-Volume, Positive Displacement Syringes |