Learn More
Hamilton™ Syringe, Cemented Needle with Guide, Sold by MilliporeSigma™ Supelco™
Microliter™ Syringes feature a stainless steel plunger that is hand fitted to its matching glass barrel.
Supplier:  MilliporeSigma Supelco 20753
 View more versions of this product
Catalog No. 11-101-1027
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. View substitute products »
Description
Description
  • Hand-fitting process creates a liquid-tight seal between plunger and barrel
  • Plungers and barrels are non-interchangeable or replaceable
  • Ideal for use with liquid handling and homogenous samples
Specifications
Specifications
Not Autoclavable
26s G
0.005 in.
0.0187 in.
0.485 mm
0.47 mm
Microliter™ Syringe
122°F
50°F
2
1 Ea.
Hamilton
Bevel
Cemented Needle with Guide
Borosilicate Glass (Barrel), Stainless Steel (Tip)
0.019 in.
0.13 mm
0.260 in.
6.60 mm
2 in.
2000 psig (137.9 bar)
50°C
701 NWG
10°C
10 μL
51 mm
Hamilton 701 NWG
Recommendations
Recommendations
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Hamilton is a registered trademark of Hamilton Co.